Learn More
Medchemexpress LLC L-Glutamic acid, 1,5-bis(1,1-dimethylethyl) ester, hydrochloride (1:1) | 32677-01-3 | 98.0% | 295.80 | 100 G
SDP
Catalog No. 5000404562
Encompass_Preferred
H-Glu(OtBu)-OtBu hydrochloride is a key intermediate utilized in the synthesis of prostate-specific membrane antigen (PSMA) targeting probes. This compound has the ability to decrease non-specific background binding through negatively charged linkers, thereby enhancing tumor/background contrast. It is applicable in prostate cancer PET/SPECT imaging studies.
  • Key intermediate for synthesizing PSMA targeting probes.
  • Reduces non-specific background binding through negatively charged linkers.
  • Improves tumor/background contrast.
  • Used in prostate cancer PET/SPECT imaging studies.
  • For research use only.
Catalog No. 50-004-04562 Supplier Medchemexpress LLC Supplier No. HY20167A100G
H-Glu(OtBu)-OtBu hydrochloride is a key intermediate utilized in the synthesis of prostate-specific membrane antigen (PSMA) targeting probes. This compound has the ability to decrease non-specific background binding through negatively charged linkers, thereby enhancing tumor/background contrast. It is applicable in prostate cancer PET/SPECT imaging studies.
  • Key intermediate for synthesizing PSMA targeting probes.
  • Reduces non-specific background binding through negatively charged linkers.
  • Improves tumor/background contrast.
  • Used in prostate cancer PET/SPECT imaging studies.
  • For research use only.