Learn More
Medchemexpress LLC L-Glutamic acid-13C2 | 115473-56-8 | >99.7% | 149.11 | 50 MG
SDP
Catalog No. 5000360259
Encompass_Preferred
L-Glutamic acid-13C2 is the 13C labeled L-Glutamic acid. It functions as an excitatory transmitter and an agonist at all subtypes of glutamate receptors, including metabotropic, kainate, NMDA, and AMPA receptors. It also exhibits a direct activating effect on the release of dopamine from dopaminergic terminals.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated for quantitation during drug development.
Catalog No. 50-003-60259 Supplier Medchemexpress LLC Supplier No. HY14608S1050MG
L-Glutamic acid-13C2 is the 13C labeled L-Glutamic acid. It functions as an excitatory transmitter and an agonist at all subtypes of glutamate receptors, including metabotropic, kainate, NMDA, and AMPA receptors. It also exhibits a direct activating effect on the release of dopamine from dopaminergic terminals.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated for quantitation during drug development.