Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamic acid-13C5,15N | 202468-31-3 | 99.9% | 13C5H915NO4 | 10MG
SDP

Catalog No. 5000259406
Encompass_Preferred

L-Glutamic acid-13C5,15N is a stable isotope-labeled form of the endogenous amino acid L-glutamic acid, containing five 13C atoms and one 15N atom. It is provided as a high-purity analytical standard for use in metabolic tracing, quantitative mass spectrometry, and metabolic flux analysis.

  • Labeled with five 13C and one 15N for clear tracer detection.
  • High purity suitable for LC-MS/MS and isotope-dilution quantitation.
  • Enables metabolic tracing and flux analysis in cellular and biochemical studies.
  • Consistent isotopic enrichment for reproducible analytical results.
  • Molecular formula: 13C5H915NO4; molecular weight: 153.09.

Catalog No. 50-002-59406 Supplier Medchemexpress LLC Supplier No. HY14608S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamic acid-13C5,15N is a stable isotope-labeled form of the endogenous amino acid L-glutamic acid, containing five 13C atoms and one 15N atom. It is provided as a high-purity analytical standard for use in metabolic tracing, quantitative mass spectrometry, and metabolic flux analysis.

  • Labeled with five 13C and one 15N for clear tracer detection.
  • High purity suitable for LC-MS/MS and isotope-dilution quantitation.
  • Enables metabolic tracing and flux analysis in cellular and biochemical studies.
  • Consistent isotopic enrichment for reproducible analytical results.
  • Molecular formula: 13C5H915NO4; molecular weight: 153.09.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.