Learn More
Medchemexpress LLC L-glutamic acid-13C5,15N | 202468-31-3 | MFCD00144596 | 99.9% | 153.09 g/mol | 13C5H9 15N O4 | 5 MG
SDP
Catalog No. 5000222838
Encompass_Preferred
L-Glutamic acid-13C5,15N is an isotopically labeled form of L-glutamic acid containing five 13C atoms and one 15N atom. Supplied as a high-purity solid, it is intended for use as a tracer or internal standard in analytical and metabolic studies, including NMR, GC-MS, and LC-MS quantitative analyses.
  • High isotopic enrichment for accurate quantitation.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity for reproducible results.
  • Supplied as a stable solid for easy handling and storage.
  • Useful for metabolic flux and labeling studies.
Catalog No. 50-002-22838 Supplier Medchemexpress LLC Supplier No. HY14608S35MG
L-Glutamic acid-13C5,15N is an isotopically labeled form of L-glutamic acid containing five 13C atoms and one 15N atom. Supplied as a high-purity solid, it is intended for use as a tracer or internal standard in analytical and metabolic studies, including NMR, GC-MS, and LC-MS quantitative analyses.
  • High isotopic enrichment for accurate quantitation.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity for reproducible results.
  • Supplied as a stable solid for easy handling and storage.
  • Useful for metabolic flux and labeling studies.