Learn More
Medchemexpress LLC L-Glutamic acid-13C5 | 55443-55-5 | 98.4% | 152.09 | 1 MG
SDP
Catalog No. 5000363509
Encompass_Preferred
L-Glutamic acid-13C5 is the 13C-labeled L-Glutamic acid. L-Glutamic acid functions as an excitatory transmitter and an agonist at all subtypes of glutamate receptors, including metabotropic, kainate, NMDA, and AMPA receptors. It also directly activates the release of dopamine (DA) from dopaminergic terminals.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development
  • Deuteration has potential to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-63509 Supplier Medchemexpress LLC Supplier No. HY14608S51MG
L-Glutamic acid-13C5 is the 13C-labeled L-Glutamic acid. L-Glutamic acid functions as an excitatory transmitter and an agonist at all subtypes of glutamate receptors, including metabotropic, kainate, NMDA, and AMPA receptors. It also directly activates the release of dopamine (DA) from dopaminergic terminals.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development
  • Deuteration has potential to influence pharmacokinetic and metabolic profiles of drugs