Learn More
Medchemexpress LLC L-glutamic acid, 4-methyl-, (4R)- | 31137-74-3 | 98.0% | 161.16 | C6H11NO4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10131050MG
Encompass_Preferred
SYM 2081 is a selective, high-affinity kainate receptor agonist used as a laboratory research reagent to study glutamatergic signaling, transporter function, neuroprotection, and apoptotic pathways. Intended for research use only and supplied as a white to off-white solid with manufacturer-provided characterization.
Catalog No. 50-002-12806