Learn More
Medchemexpress LLC L-glutamic acid, 4-methyl-, (4R)- | 31137-74-3 | MFCD00937765 | 98.0% | 161.16 | C6H11NO4 | 10 MG
SDP
Catalog No. 5000212650
Encompass_Preferred
SYM 2081 is a selective kainate receptor agonist used as a research reagent to investigate glutamate receptor function, excitatory neurotransmission, and neuroprotective signaling. It has documented activity in cellular and animal models of hypoxic-ischemic injury and inflammatory or neuropathic pain, and affects glutamate transport mechanisms.
  • Selective kainate receptor agonist for receptor pharmacology studies.
  • Modulates glutamate transport (EAAT1 substrate, EAAT2 inhibitor).
  • Demonstrates neuroprotective effects in cellular and animal models.
  • High purity suitable for research applications.
  • Supplied in small milligram quantities with defined storage conditions.
Catalog No. 50-002-12650 Supplier Medchemexpress LLC Supplier No. HY10131010MG
SYM 2081 is a selective kainate receptor agonist used as a research reagent to investigate glutamate receptor function, excitatory neurotransmission, and neuroprotective signaling. It has documented activity in cellular and animal models of hypoxic-ischemic injury and inflammatory or neuropathic pain, and affects glutamate transport mechanisms.
  • Selective kainate receptor agonist for receptor pharmacology studies.
  • Modulates glutamate transport (EAAT1 substrate, EAAT2 inhibitor).
  • Demonstrates neuroprotective effects in cellular and animal models.
  • High purity suitable for research applications.
  • Supplied in small milligram quantities with defined storage conditions.