Learn More
Medchemexpress LLC L-glutamic acid, 4-methyl-, (4R)- | 31137-74-3 | MFCD00937765 | 98.0% | 161.16 g/mol | C6H11NO4 | 25 MG
SDP
Catalog No. 5000211129
Encompass_Preferred
SYM 2081 is a research-grade kainate receptor agonist (2S,4R-4-methylglutamic acid) used in neuropharmacology to probe kainate-type glutamate receptors and excitatory amino acid transporter activity. Supplied as a powder for laboratory research; not for human use.
  • Potent kainate receptor agonist for receptor pharmacology studies.
  • High purity: 98.0%.
  • Molecular formula C6H11NO4; molecular weight 161.16 g/mol.
  • Powder form with storage recommendations: -20°C (3 years) or 4°C (2 years).
  • Available in small research quantities, including 25 mg size.
Catalog No. 50-002-11129 Supplier Medchemexpress LLC Supplier No. HY10131025MG
SYM 2081 is a research-grade kainate receptor agonist (2S,4R-4-methylglutamic acid) used in neuropharmacology to probe kainate-type glutamate receptors and excitatory amino acid transporter activity. Supplied as a powder for laboratory research; not for human use.
  • Potent kainate receptor agonist for receptor pharmacology studies.
  • High purity: 98.0%.
  • Molecular formula C6H11NO4; molecular weight 161.16 g/mol.
  • Powder form with storage recommendations: -20°C (3 years) or 4°C (2 years).
  • Available in small research quantities, including 25 mg size.