Learn More
Research Products International Corp L-Glutamic Acid, 500 Grams
SDP
Catalog No. 50213337
Encompass_Preferred
L-Glutamic Acid is used in cell culture media for the cultivation of various cell types. It serves as a source of carbon and nitrogen for cell growth. 
Also, is a common component in laboratory research, particularly in studies related to neuroscience, biochemistry, and molecular biology. 
Its role as an amino acid and neurotransmitter makes it valuable for understanding cellular and neural processes.
Catalog No. 50-213-337 Supplier Research Products International Corp Supplier No. G36020500.0
L-Glutamic Acid is used in cell culture media for the cultivation of various cell types. It serves as a source of carbon and nitrogen for cell growth. 
Also, is a common component in laboratory research, particularly in studies related to neuroscience, biochemistry, and molecular biology. 
Its role as an amino acid and neurotransmitter makes it valuable for understanding cellular and neural processes.