Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Glutamic acid, compd. with L-arginine (1:1) | 4320-30-3 | 321.33 | 1 G
SDP

Catalog No. 5000389707
Encompass_Preferred

L-Arginine L-glutamate ((S)-(+)-Arginine L-glutamate) is a nitrogen donor for the synthesis of nitric oxide. It can be used for research related to upper gastrointestinal hypofunction or dysfunction, such as functional dyspepsia.

  • Nitrogen donor for synthesis of nitric oxide
  • Can be used for research on upper gastrointestinal hypofunction or dysfunction (e.g., functional dyspepsia)
  • Dose-dependently promotes gastric emptying in rats, a vagus nerve-dependent effect
  • Dose-dependently enhances adaptive relaxation of rat stomachs
  • Purity: 98.0%
  • Appearance: Solid, white to off-white
  • Initial source: Endogenous metabolite

Catalog No. 50-003-89707 Supplier Medchemexpress LLC Supplier No. HYN0455B1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Arginine L-glutamate ((S)-(+)-Arginine L-glutamate) is a nitrogen donor for the synthesis of nitric oxide. It can be used for research related to upper gastrointestinal hypofunction or dysfunction, such as functional dyspepsia.

  • Nitrogen donor for synthesis of nitric oxide
  • Can be used for research on upper gastrointestinal hypofunction or dysfunction (e.g., functional dyspepsia)
  • Dose-dependently promotes gastric emptying in rats, a vagus nerve-dependent effect
  • Dose-dependently enhances adaptive relaxation of rat stomachs
  • Purity: 98.0%
  • Appearance: Solid, white to off-white
  • Initial source: Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.