Learn More
Medchemexpress LLC L-glutamic acid-d5 | 2784-50-1 | MFCD00144590 | 99.8% | 152.16 g/mol | C5H4D5NO4 | 5 MG
SDP
Catalog No. 5000221269
Encompass_Preferred
L-Glutamic acid-d5 is the deuterium-labeled form of L-glutamic acid supplied for analytical and biochemical research. It is intended for use as a stable isotope reagent and tracer in quantitative experiments, offering reliable performance in mass spectrometry and metabolic studies.
  • Suitable as an internal standard for LC-MS and GC-MS assays.
  • Supports metabolic profiling and tracer experiments with minimal isotopic interference.
  • High isotopic and chemical purity: 99.8%.
  • Available in small, research-scale quantities for analytical workflows.
  • Stable when stored as a powder at recommended low temperatures.
Catalog No. 50-002-21269 Supplier Medchemexpress LLC Supplier No. HY14608S75MG
L-Glutamic acid-d5 is the deuterium-labeled form of L-glutamic acid supplied for analytical and biochemical research. It is intended for use as a stable isotope reagent and tracer in quantitative experiments, offering reliable performance in mass spectrometry and metabolic studies.
  • Suitable as an internal standard for LC-MS and GC-MS assays.
  • Supports metabolic profiling and tracer experiments with minimal isotopic interference.
  • High isotopic and chemical purity: 99.8%.
  • Available in small, research-scale quantities for analytical workflows.
  • Stable when stored as a powder at recommended low temperatures.