Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid dibenzyl ester hydrochloride, 98%

Catalog No. AAH63598MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63598MD 250 mg
AAH6359803 1 g
AAH6359806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63598MD Supplier Thermo Scientific Chemicals Supplier No. H63598MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4561-10-8 | C19H22ClNO4 | 363.84 g/mol

L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4561-10-8
MDL Number MFCD00035103

Specifications

Color White
Molecular Formula C17H38ClN
Synonym tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride
InChI Key CEYYIKYYFSTQRU-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-]
IUPAC Name trimethyl(tetradecyl)azanium;chloride
Quantity 250 mg
Molecular Weight (g/mol) 291.95
PubChem CID 20708
Formula Weight 363.84
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Glutamic acid dibenzyl ester hydrochloride
Show More Show Less
RTECSNumber MA0885000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.