Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid dibenzyl ester p-toluenesulfonate, 98%, Thermo Scientific™

Catalog No. AAH6269714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6269714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6269714 Supplier Thermo Scientific Chemicals Supplier No. H6269714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2791-84-6
Molecular Formula C26H29NO7S
Molecular Weight (g/mol) 499.578
MDL Number MFCD00038980
InChI Key HVZUAIVKRYGQRM-LMOVPXPDSA-N
Synonym h-glu obzl-obzl.tosoh, l-glutamic acid dibenzyl ester tosylate, s-dibenzyl 2-aminopentanedioate 4-methylbenzenesulfonate, h-glu obzl-obzl p-tosylate, l-glutamic acid dibenzyl ester 4-toluenesulfonate, glu obzl-obzl.tosoh, dibenzyl l-glutamate p-toluenesulfonate, h-glu obzl-obzl 4-tosylate, l-glutamic acid dibenzyl ester p-toluenesulfonate, h-glu obzl-obzl inverted exclamation mark currencytosoh
PubChem CID 13233366
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N

Specifications

Color White
Quantity 25 g
Formula Weight 499.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Glutamic acid dibenzyl ester p-toluenesulfonate
EINECSNumber 220-522-4
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.