Learn More
Medchemexpress LLC L-glutamic acid gamma-(7-amido-4-methylcoumarin) | 98516-76-8 | MFCD00051217 | >98.0% | 304.30 g·mol⁻¹ | C15H16N2O5 | 100 MG
SDP
Catalog No. 5000281703
Encompass_Preferred
H-Glu(AMC)-OH is a fluorescent amino acid derivative used as a peptide substrate to measure aminopeptidase A activity. The compound contains a 7-amino-4-methylcoumarin (AMC) fluorophore linked to glutamic acid; enzymatic cleavage releases the fluorescent AMC reporter for sensitive detection. Supplied as a solid for research use, it is suitable for kinetic and end-point enzymology assays.
Catalog No. 50-002-81703 Supplier Medchemexpress LLC Supplier No. HYW250725100MG
H-Glu(AMC)-OH is a fluorescent amino acid derivative used as a peptide substrate to measure aminopeptidase A activity. The compound contains a 7-amino-4-methylcoumarin (AMC) fluorophore linked to glutamic acid; enzymatic cleavage releases the fluorescent AMC reporter for sensitive detection. Supplied as a solid for research use, it is suitable for kinetic and end-point enzymology assays.