Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamic acid, homopolymer, sodium | 26247-79-0 | MFCD00081868 | 30,000 Da (average) | (C5H6NO3Na)n | 10 MG
SDP

Catalog No. 5000284057
Encompass_Preferred

Poly-L-glutamic acid sodium salt (average MW ~30,000 Da) is a water-soluble polypeptide supplied as an off-white to light yellow solid for research use. It is commonly used as a negatively charged polymer in biochemical assays, conjugation chemistry, and biomaterials research.

  • Sodium salt form of poly-L-glutamic acid.
  • Average molecular weight ~30,000 Da.
  • CAS number 26247-79-0.
  • Off-white to light yellow solid physical appearance.
  • Available in small quantities (e.g., 10 MG).
  • Recommended storage: protect from light; long-term at -20°C; in solution: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-84057 Supplier Medchemexpress LLC Supplier No. HYW596474D10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly-L-glutamic acid sodium salt (average MW ~30,000 Da) is a water-soluble polypeptide supplied as an off-white to light yellow solid for research use. It is commonly used as a negatively charged polymer in biochemical assays, conjugation chemistry, and biomaterials research.

  • Sodium salt form of poly-L-glutamic acid.
  • Average molecular weight ~30,000 Da.
  • CAS number 26247-79-0.
  • Off-white to light yellow solid physical appearance.
  • Available in small quantities (e.g., 10 MG).
  • Recommended storage: protect from light; long-term at -20°C; in solution: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.