Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamic acid, homopolymer, sodium salt | 26247-79-0 | MFCD00081868 | >98.0% | 30,000 Da (average) | (C5H9NO4)x · xNa | 5 MG
SDP

Catalog No. 5000284526
Encompass_Preferred

Poly-L-glutamic acid sodium salt (average molecular weight 30,000 Da) is a water-soluble polypeptide supplied as an off-white to light-yellow solid for research use. It functions as a charged, biodegradable polymer used in biochemical assays and drug-delivery research; it is soluble in water and supplied in small research-scale quantities.

  • High-molecular-weight sodium salt of poly-L-glutamic acid (avg MW ~30,000 Da)
  • Off-white to light-yellow solid, water soluble (≈125 mg/mL; ultrasonic treatment may be required)
  • Intended for research use in biochemical assays and delivery studies
  • CAS number 26247-79-0; polymeric repeating unit (C5H9NO4)x · xNa
  • Storage: -20°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-84526 Supplier Medchemexpress LLC Supplier No. HYW596474D5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly-L-glutamic acid sodium salt (average molecular weight 30,000 Da) is a water-soluble polypeptide supplied as an off-white to light-yellow solid for research use. It functions as a charged, biodegradable polymer used in biochemical assays and drug-delivery research; it is soluble in water and supplied in small research-scale quantities.

  • High-molecular-weight sodium salt of poly-L-glutamic acid (avg MW ~30,000 Da)
  • Off-white to light-yellow solid, water soluble (≈125 mg/mL; ultrasonic treatment may be required)
  • Intended for research use in biochemical assays and delivery studies
  • CAS number 26247-79-0; polymeric repeating unit (C5H9NO4)x · xNa
  • Storage: -20°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.