Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamic acid monopotassium salt | 19473-49-5 | MFCD00035545 | 98.0% | 185.22 | C5H8KNO4 | 5 MG
SDP

Catalog No. 5000282083
Encompass_Preferred

L-Glutamic acid monopotassium salt is the monopotassium salt of the amino acid L-glutamic acid. It is supplied for research use and described as a food additive and flavor enhancer that can reduce the harmful effects of lithium. The compound has formula C5H8KNO4 and a molecular weight of 185.22 g/mol.

  • Potassium salt of L-glutamic acid
  • Food additive and flavor enhancer
  • Reported to reduce harmful effects of lithium
  • Purity 98.0%
  • Molecular weight 185.22 g/mol
  • Available in small research quantities, including 5 mg

Catalog No. 50-002-82083 Supplier Medchemexpress LLC Supplier No. HYW3377395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamic acid monopotassium salt is the monopotassium salt of the amino acid L-glutamic acid. It is supplied for research use and described as a food additive and flavor enhancer that can reduce the harmful effects of lithium. The compound has formula C5H8KNO4 and a molecular weight of 185.22 g/mol.

  • Potassium salt of L-glutamic acid
  • Food additive and flavor enhancer
  • Reported to reduce harmful effects of lithium
  • Purity 98.0%
  • Molecular weight 185.22 g/mol
  • Available in small research quantities, including 5 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.