Learn More
Medchemexpress LLC L-glutamic acid monopotassium salt | 19473-49-5 | MFCD03938965 | 185.22 g/mol | C5H8KNO4 | 25 MG
SDP
Catalog No. 5000282389
Encompass_Preferred
L-Glutamic acid monopotassium salt is the potassium salt of L-glutamic acid supplied as a white to off-white solid for research use. It is commonly used as a biochemical reagent and flavoring agent in formulation studies and biochemical assays, provided in small milligram pack sizes for bench-scale experiments.
  • Potassium salt of L-glutamic acid used as a biochemical reagent.
  • White to off-white solid form suitable for laboratory handling.
  • Common applications include flavor enhancer studies and biochemical assays.
  • Available in small milligram pack sizes for bench-scale research.
  • Includes CAS number 19473-49-5 and molecular weight 185.22 g/mol for unambiguous identification.
  • Store dry and cool; in solution, store at -20°C (1 year) or -80°C (2 years).
Catalog No. 50-002-82389 Supplier Medchemexpress LLC Supplier No. HYW33773925MG
L-Glutamic acid monopotassium salt is the potassium salt of L-glutamic acid supplied as a white to off-white solid for research use. It is commonly used as a biochemical reagent and flavoring agent in formulation studies and biochemical assays, provided in small milligram pack sizes for bench-scale experiments.
  • Potassium salt of L-glutamic acid used as a biochemical reagent.
  • White to off-white solid form suitable for laboratory handling.
  • Common applications include flavor enhancer studies and biochemical assays.
  • Available in small milligram pack sizes for bench-scale research.
  • Includes CAS number 19473-49-5 and molecular weight 185.22 g/mol for unambiguous identification.
  • Store dry and cool; in solution, store at -20°C (1 year) or -80°C (2 years).