Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamic acid (monopotassium salt) | 19473-49-5 | MFCD23135485 | 98.0% | 185.22 g/mol | C5H8KNO4 | 10 MG
SDP

Catalog No. 5000282388
Encompass_Preferred

L-Glutamic acid monopotassium salt is the monopotassium salt of the amino acid L-glutamic acid. It is supplied as a white, water-soluble crystalline reagent used as a food additive and flavor enhancer and for biochemical research and applications.

  • High purity: 98.0%.
  • CAS number: 19473-49-5.
  • Molecular weight: 185.22 g/mol.
  • Water soluble solid suitable for reagent use.
  • Available in small packaged quantities, including 10 mg, for laboratory studies.

Catalog No. 50-002-82388 Supplier Medchemexpress LLC Supplier No. HYW33773910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamic acid monopotassium salt is the monopotassium salt of the amino acid L-glutamic acid. It is supplied as a white, water-soluble crystalline reagent used as a food additive and flavor enhancer and for biochemical research and applications.

  • High purity: 98.0%.
  • CAS number: 19473-49-5.
  • Molecular weight: 185.22 g/mol.
  • Water soluble solid suitable for reagent use.
  • Available in small packaged quantities, including 10 mg, for laboratory studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.