Learn More
Medchemexpress LLC HY-14608A 100mg Medchemexpress, L-Glutamic acid monosodium salt CAS:142-47-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14608A100MG
Encompass_Preferred
Medchemexpress, HY-14608A 100mg L-Glutamic acid monosodium salt CAS:142-47-2 L-Glutamic acid monosodium salt acts as an excitatory transmitter and an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA). (S)-Glutamic acid shows a direct activating effect on the release of DA from dopaminergic terminals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1624