Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid monosodium salt hydrate, ≥98.5% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111013097 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3097 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3097 Supplier MilliporeSigma Supelco Supplier No. 014951G
Add to Cart
Add to Cart

Specifications

CAS 142-47-2 (anhydrous)
Linear Formula C5H8 NNaO4 · xH2 O
MDL Number MFCD00013074
Synonym (S)-2-Aminopentanedioic acid; L-Glutamic acid monosodium salt; Glu; MSG; Monosodium glutamate
Type Analytical Standard
Quantity 1 g
Formula Weight 169.11 (anhydrous basis)
Percent Purity ≥98.5% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.