Learn More
Medchemexpress LLC L-Glutamic acid, N-[(1,1-dimethylethoxy)carbonyl]-, 5-(1,1-dimethylethyl) ester | 13726-84-6 | 98.0% | 303.35 | 25 G
SDP
Catalog No. 5000451491
Encompass_Preferred
Boc-Glu(OtBu)-OH is a protected glutamic acid derivative with two protecting groups: Boc- and OTBu. It can be used in the study of peptide compound synthesis.
  • Protected glutamic acid derivative
  • Contains Boc- and OTBu protecting groups
  • Used in peptide compound synthesis
Catalog No. 50-004-51491 Supplier Medchemexpress LLC Supplier No. 25GHYW009229
Boc-Glu(OtBu)-OH is a protected glutamic acid derivative with two protecting groups: Boc- and OTBu. It can be used in the study of peptide compound synthesis.
  • Protected glutamic acid derivative
  • Contains Boc- and OTBu protecting groups
  • Used in peptide compound synthesis