Learn More
Medchemexpress LLC L-Glutamic acid, N-[(1,1-dimethylethoxy)carbonyl]-, 5-(phenylmethyl) ester | 13574-13-5 | 95.0% | 337.37 | 100 G
SDP
Catalog No. 5000454702
Encompass_Preferred
Boc-Glu(OBzl)-OH (Compound 9) is a glutamic acid derivative commonly used in Boc Solid Phase Peptide Synthesis (SPPS). Glutamic acid residues increase the hydrophilicity of the polypeptide and play an important structural and receptor binding role.
  • Glutamic acid derivative
  • Commonly used in Boc SPPS
  • Glutamic acid residues increase polypeptide hydrophilicity
  • Plays important structural and receptor binding role
Catalog No. 50-004-54702 Supplier Medchemexpress LLC Supplier No. HYW013781100G
Boc-Glu(OBzl)-OH (Compound 9) is a glutamic acid derivative commonly used in Boc Solid Phase Peptide Synthesis (SPPS). Glutamic acid residues increase the hydrophilicity of the polypeptide and play an important structural and receptor binding role.
  • Glutamic acid derivative
  • Commonly used in Boc SPPS
  • Glutamic acid residues increase polypeptide hydrophilicity
  • Plays important structural and receptor binding role