Learn More
Medchemexpress LLC L-Glutamic acid, N-[4-[[(2-amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl)amino]benzoyl]- | 59-30-3 | 99.0% | 441.40 | 100 MG
SDP
Catalog No. 5000392393
Encompass_Preferred
Folic acid (Vitamin B9) is an orally active essential nutrient from the B complex group of vitamins. It demonstrates an antidepressant-like effect and its sodium form can reduce the risk of neonatal neural tube defects. Folic acid is also used in the research of megaloblastic and macrocytic anemias caused by folic deficiency.
  • Exhibits an antidepressant-like effect.
  • Sodium form reduces the risk of neonatal neural tube defects.
  • Used in research of megaloblastic and macrocytic anemias due to folic deficiency.
  • Plays a critical role in preventing chromosome breakage and hypomethylation of DNA.
  • Can be used to induce acute kidney injury models.
Catalog No. 50-003-92393 Supplier Medchemexpress LLC Supplier No. HY16637100MG
Folic acid (Vitamin B9) is an orally active essential nutrient from the B complex group of vitamins. It demonstrates an antidepressant-like effect and its sodium form can reduce the risk of neonatal neural tube defects. Folic acid is also used in the research of megaloblastic and macrocytic anemias caused by folic deficiency.
  • Exhibits an antidepressant-like effect.
  • Sodium form reduces the risk of neonatal neural tube defects.
  • Used in research of megaloblastic and macrocytic anemias due to folic deficiency.
  • Plays a critical role in preventing chromosome breakage and hypomethylation of DNA.
  • Can be used to induce acute kidney injury models.