Learn More
Cayman Chemical L-Glutamic Acid γ-p-Nitroanilide (hydrate), Purity: ≥95%, CAS: 122864-94-2, 5 g
SDP
Catalog No. 502912283
Encompass_Preferred
A colorimetric substrate for γ-glutamyl transpeptidase; pNA is released upon enzymatic cleavage which can be quantified by colorimetric detection at 405 nm as a measure of γ-glutamyl transpeptidase activity
Catalog No. 50-291-2283 Supplier Cayman Chemical Supplier No. 362095 G
A colorimetric substrate for γ-glutamyl transpeptidase; pNA is released upon enzymatic cleavage which can be quantified by colorimetric detection at 405 nm as a measure of γ-glutamyl transpeptidase activity