Learn More
Medchemexpress LLC L-Glutamic acid, polymer with L-alanine, L-lysine and L-tyrosine, acetate (salt) | 147245-92-9 | 98.0% | 200 MG
SDP
Catalog No. 5000345521
Encompass_Preferred
Glatiramer acetate is a synthetic analogue of myelin basic protein and an immunomodulating agent. It inhibits Experimental autoimmune encephalomyelitis (EAE) and can be used for research on multiple sclerosis. It binds strongly and promiscuously to MHC molecules, competing with various myelin antigens for presentation to T cells. Additionally, it potently induces specific suppressor cells of the T helper 2 (Th2) type, which migrate to the brain and lead to in situ bystander suppression.
  • Immunomodulating agent.
  • Inhibits Experimental autoimmune encephalomyelitis (EAE).
  • Can be used for the research of multiple sclerosis.
  • Exhibits strong and promiscuous binding to MHC molecules.
  • Induces specific suppressor cells of the T helper 2 (Th2) type.
  • Increases BDNF levels (in vitro studies).
  • Improves performance in motor function measures in Huntington's disease mouse models (in vivo studies).
Catalog No. 50-003-45521 Supplier Medchemexpress LLC Supplier No. HY109520100MG
Glatiramer acetate is a synthetic analogue of myelin basic protein and an immunomodulating agent. It inhibits Experimental autoimmune encephalomyelitis (EAE) and can be used for research on multiple sclerosis. It binds strongly and promiscuously to MHC molecules, competing with various myelin antigens for presentation to T cells. Additionally, it potently induces specific suppressor cells of the T helper 2 (Th2) type, which migrate to the brain and lead to in situ bystander suppression.
  • Immunomodulating agent.
  • Inhibits Experimental autoimmune encephalomyelitis (EAE).
  • Can be used for the research of multiple sclerosis.
  • Exhibits strong and promiscuous binding to MHC molecules.
  • Induces specific suppressor cells of the T helper 2 (Th2) type.
  • Increases BDNF levels (in vitro studies).
  • Improves performance in motor function measures in Huntington's disease mouse models (in vivo studies).