Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Glutamic acid, polymer with L-alanine, L-lysine and L-tyrosine, acetate (salt) | 147245-92-9 | 98.0% | 25 MG
SDP

Catalog No. 5000347563
Encompass_Preferred

L-Glutamic acid, polymer with L-alanine, L-lysine and L-tyrosine, acetate (salt) | 147245-92-9 | 98.0% | 25 MG

Catalog No. 50-003-47563 Supplier Medchemexpress LLC Supplier No. 25MGHY109520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamic acid, polymer with L-alanine, L-lysine and L-tyrosine, acetate (salt) | 147245-92-9 | 98.0% | 25 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.