Learn More
Alkali Scientific L Glutamic Acid Potassium Salt Monohydrate, 100 G
SDP
Catalog No. 5000661100
Encompass_Preferred
L Glutamic Acid Potassium Salt Monohydrate, 100 G, is a high-purity amino acid reagent used in biochemical research and in the preparation of specialized culture media. This salt form of glutamic acid is commonly used as a nitrogen source in microbiological media and as a component in buffer solutions. It is often used in the synthesis of peptides and proteins, particularly in the study of neurotransmitters, metabolism, and enzymatic reactions. As a vital nutrient for various organisms, this potassium salt form of glutamic acid ensures stable pH levels in solutions and supports cell growth in research applications. It meets stringent biochemical reagent standards and is suitable for laboratory use.
Catalog No. 50-006-61100 Supplier Alkali Scientific Supplier No. LGL003CP
L Glutamic Acid Potassium Salt Monohydrate, 100 G, is a high-purity amino acid reagent used in biochemical research and in the preparation of specialized culture media. This salt form of glutamic acid is commonly used as a nitrogen source in microbiological media and as a component in buffer solutions. It is often used in the synthesis of peptides and proteins, particularly in the study of neurotransmitters, metabolism, and enzymatic reactions. As a vital nutrient for various organisms, this potassium salt form of glutamic acid ensures stable pH levels in solutions and supports cell growth in research applications. It meets stringent biochemical reagent standards and is suitable for laboratory use.