Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamic acid potassium salt monohydrate | 6382-01-0 | MFCD00150343 | 98.0% | 203.23 g/mol | C5H10KNO5 | 10 G
SDP

Catalog No. 5000283861
Encompass_Preferred

L-Glutamic acid potassium salt monohydrate is the monopotassium salt and monohydrate form of the amino acid L-glutamic acid. Supplied for research use, it serves as an amino acid source and buffering component in biochemical, cell culture, and analytical applications. Common uses include media formulation, enzymatic assays, and analytical reference work.

  • Purity 98.0%.
  • Molecular weight 203.23 g/mol.
  • Soluble in water for easy formulation.
  • Available in small lab pack sizes for bench-scale experiments.
  • Suitable for biochemical assays and cell culture media formulation.
  • Provided with SDS and certificate of analysis for quality assurance.

Catalog No. 50-002-83861 Supplier Medchemexpress LLC Supplier No. HYW14194910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamic acid potassium salt monohydrate is the monopotassium salt and monohydrate form of the amino acid L-glutamic acid. Supplied for research use, it serves as an amino acid source and buffering component in biochemical, cell culture, and analytical applications. Common uses include media formulation, enzymatic assays, and analytical reference work.

  • Purity 98.0%.
  • Molecular weight 203.23 g/mol.
  • Soluble in water for easy formulation.
  • Available in small lab pack sizes for bench-scale experiments.
  • Suitable for biochemical assays and cell culture media formulation.
  • Provided with SDS and certificate of analysis for quality assurance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.