Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamic acid potassium salt monohydrate | 6382-01-0 | MFCD00150343 | 98.0% | 203.23 g/mol | C5H10KNO5 | 5 G
SDP

Catalog No. 5000283537
Encompass_Preferred

L-glutamic acid potassium salt monohydrate is the potassium salt, monohydrate form of the amino acid L-glutamic acid supplied as a purified solid for laboratory research. It is used in biochemical assays, buffer formulation, and cell culture where a potassium counter-ion or hydrated form is required.

  • Purity 98.0%.
  • Molecular weight 203.23 g/mol.
  • Chemical formula C5H10KNO5.
  • Provided as a dry powder suitable for dissolution and formulation.
  • Available in small pack sizes for laboratory use.
  • Useful as a biochemical reagent, buffer component, or nutrient source.

Catalog No. 50-002-83537 Supplier Medchemexpress LLC Supplier No. HYW1419495G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-glutamic acid potassium salt monohydrate is the potassium salt, monohydrate form of the amino acid L-glutamic acid supplied as a purified solid for laboratory research. It is used in biochemical assays, buffer formulation, and cell culture where a potassium counter-ion or hydrated form is required.

  • Purity 98.0%.
  • Molecular weight 203.23 g/mol.
  • Chemical formula C5H10KNO5.
  • Provided as a dry powder suitable for dissolution and formulation.
  • Available in small pack sizes for laboratory use.
  • Useful as a biochemical reagent, buffer component, or nutrient source.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.