Learn More
Bioworld L-GLUTAMIC ACID SEPAROPORE
Supplier:  Bioworld 201810683
Encompass
L-GLUTAMIC ACID SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: L-Glutamic Acid.; Particle size (range): 50-175m.; Molecular Weight range: 6x104 - 2x107.; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads).; pH stability: 4-9.; ...
Catalog No. 50-197-842