Learn More
Medchemexpress LLC L-glutamic acid, sodium salt, hydrate | 6106-04-3 | MFCD00150138 | 98.0% | 187.13 | C5H10NNaO5 | 5 G
SDP
Catalog No. 5000302665
Encompass_Preferred
L-Glutamic acid monosodium (hydrate) is the monohydrate sodium salt of L-glutamic acid supplied as a white to off-white solid for laboratory and research use. It functions as a biochemical reagent, nutritional additive/flavoring agent, and can be used to induce oxidative stress in cell models. Typical specs include CAS 6106-04-3, molecular weight 187.13, formula C5H10NNaO5, and purity around 98.0%.
  • High purity reagent (98.0%).
  • White to off-white solid, easy to weigh and store.
  • Soluble in water: ~100 mg/mL; ultrasonic agitation recommended.
  • Suitable as a biochemical reagent and flavoring additive.
  • Can be used to induce oxidative stress in cellular assays.
  • Stable when stored dry at room temperature; short-term stability in solution at frozen temperatures.
Catalog No. 50-003-02665 Supplier Medchemexpress LLC Supplier No. HYW0161455G
L-Glutamic acid monosodium (hydrate) is the monohydrate sodium salt of L-glutamic acid supplied as a white to off-white solid for laboratory and research use. It functions as a biochemical reagent, nutritional additive/flavoring agent, and can be used to induce oxidative stress in cell models. Typical specs include CAS 6106-04-3, molecular weight 187.13, formula C5H10NNaO5, and purity around 98.0%.
  • High purity reagent (98.0%).
  • White to off-white solid, easy to weigh and store.
  • Soluble in water: ~100 mg/mL; ultrasonic agitation recommended.
  • Suitable as a biochemical reagent and flavoring additive.
  • Can be used to induce oxidative stress in cellular assays.
  • Stable when stored dry at room temperature; short-term stability in solution at frozen temperatures.