Learn More
Medchemexpress LLC L-Glutamic acid (Standard) | 56-86-0 | 99.9% | 147.13 | 50 MG
SDP
Catalog No. 5000363510
Encompass_Preferred
L-Glutamic acid (Standard) is an analytical standard intended for research and analytical applications. It functions as an excitatory amino acid neurotransmitter, acting as an agonist for all subtypes of glutamate receptors. This compound can also affect the release of DA from dopaminergic nerve endings, making it useful in the study of neurological diseases.
  • Serves as a reference standard for assays and research experiments
  • Functions as an agonist for all glutamate receptor subtypes
  • Applicable in studies related to neurological diseases
  • Appears as a solid, white to off-white substance
Catalog No. 50-003-63510 Supplier Medchemexpress LLC Supplier No. HY14608R50MG
L-Glutamic acid (Standard) is an analytical standard intended for research and analytical applications. It functions as an excitatory amino acid neurotransmitter, acting as an agonist for all subtypes of glutamate receptors. This compound can also affect the release of DA from dopaminergic nerve endings, making it useful in the study of neurological diseases.
  • Serves as a reference standard for assays and research experiments
  • Functions as an agonist for all glutamate receptor subtypes
  • Applicable in studies related to neurological diseases
  • Appears as a solid, white to off-white substance