Learn More
Cayman Chemical L-Glutamic Acid (ammonium salt), Purity: ≥95%, CAS: 7558-63-6, 5 g
SDP
Catalog No. 502992652
Encompass_Preferred
An excitatory neurotransmitter; acts on ionotropic receptors and metabotropic receptors to induce excitatory synaptic transmission and has roles in synaptic plasticity; excessive release induces excitotoxicity that is associated with various human diseases; excessive release of the protonated glutamate form occurs during seizure activity and contributes to epileptogenesis and seizure-induced brain damage
This item is not returnable. View return policy
Catalog No. 50-299-2652 Supplier Cayman Chemical Supplier No. 303775 G
An excitatory neurotransmitter; acts on ionotropic receptors and metabotropic receptors to induce excitatory synaptic transmission and has roles in synaptic plasticity; excessive release induces excitotoxicity that is associated with various human diseases; excessive release of the protonated glutamate form occurs during seizure activity and contributes to epileptogenesis and seizure-induced brain damage