Learn More
Medchemexpress LLC L-glutamic γ-monohydroxamate | 1955-67-5 | MFCD00057719 | >97.0% | C5H10N2O4 | 10MG
SDP
Catalog No. 5000259008
Encompass_Preferred
L-Glutamic γ-monohydroxamate is an amino acid-derived hydroxamate used as a research reagent in biochemical and pharmacological studies. It has been reported to inhibit cell proliferation and is investigated for antitumor activity and metal coordination chemistry.
  • Used in biochemical and pharmacological research
  • Reported to inhibit cell proliferation and exhibit antitumor activity
  • Molecular formula C5H10N2O4 and molecular weight 162.14 g/mol
  • Typically supplied in small research quantities suitable for laboratory testing
  • Typical supplier-reported purity ≥97% (method may vary)
Catalog No. 50-002-59008 Supplier Medchemexpress LLC Supplier No. HY13787410MG
L-Glutamic γ-monohydroxamate is an amino acid-derived hydroxamate used as a research reagent in biochemical and pharmacological studies. It has been reported to inhibit cell proliferation and is investigated for antitumor activity and metal coordination chemistry.
  • Used in biochemical and pharmacological research
  • Reported to inhibit cell proliferation and exhibit antitumor activity
  • Molecular formula C5H10N2O4 and molecular weight 162.14 g/mol
  • Typically supplied in small research quantities suitable for laboratory testing
  • Typical supplier-reported purity ≥97% (method may vary)