Learn More
Cayman Chemical L-GlutamIn 500mg
SDP
Catalog No. 502994253
Encompass_Preferred
A conditionally essential amino acid; involved in many biochemical processes; synthesized in vivo by glutamate and ammonia; a substrate for the biosynthesis of glutamate and GABA in neurons; decreases adhesion of sickle RBCs to HUVECs when incubated ex vivo with patient-derived autologous plasma either alone or with LPS; commonly used in cell culture media
Catalog No. 50-299-4253 Supplier Cayman Chemical Supplier No. 23716500 MG
A conditionally essential amino acid; involved in many biochemical processes; synthesized in vivo by glutamate and ammonia; a substrate for the biosynthesis of glutamate and GABA in neurons; decreases adhesion of sickle RBCs to HUVECs when incubated ex vivo with patient-derived autologous plasma either alone or with LPS; commonly used in cell culture media