Learn More
Cayman Chemical L-Glutamine-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 14341-78-7, 10 mg
SDP
Catalog No. 502912248
Encompass_Preferred
An internal standard for the quantification of L-glutamine by GC- or LC-MS
Catalog No. 50-291-2248 Supplier Cayman Chemical Supplier No. 3483910 MG
An internal standard for the quantification of L-glutamine by GC- or LC-MS