Learn More
Medchemexpress LLC L-Glutamine-1-13C (L-Glutamic acid 5-amide-1-13C) | 159663-16-8 | 147.14 | 1 MG
SDP
Catalog No. 5000390084
Encompass_Preferred
L-Glutamine-1-13C is the 13C-labeled L-Glutamine. L-Glutamine, also known as L-Glutamic acid 5-amide, is a non-essential amino acid abundant throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can be incorporated into drug molecules as tracers for quantitation during drug development.
Catalog No. 50-003-90084 Supplier Medchemexpress LLC Supplier No. HYN0390S51MG
L-Glutamine-1-13C is the 13C-labeled L-Glutamine. L-Glutamine, also known as L-Glutamic acid 5-amide, is a non-essential amino acid abundant throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can be incorporated into drug molecules as tracers for quantitation during drug development.