Learn More
Medchemexpress LLC L-Glutamine-1,2-13C2 | 56-85-9 | 148.13 | 5 MG
SDP
Catalog No. 5000388833
Encompass_Preferred
L-Glutamine-1,2-13C2 is the 13C-labeled version of L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is an abundant non-essential amino acid in the body, participating in numerous metabolic processes and providing a carbon source for oxidation in certain cells.
  • 13C-labeled L-Glutamine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Non-essential amino acid involved in many metabolic processes.
  • Provides carbons for oxidation in some cells.
Catalog No. 50-003-88833 Supplier Medchemexpress LLC Supplier No. HYN0390S105MG
L-Glutamine-1,2-13C2 is the 13C-labeled version of L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is an abundant non-essential amino acid in the body, participating in numerous metabolic processes and providing a carbon source for oxidation in certain cells.
  • 13C-labeled L-Glutamine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Non-essential amino acid involved in many metabolic processes.
  • Provides carbons for oxidation in some cells.