Learn More
Medchemexpress LLC L-glutamine-1,2-13C2 (L-glutamic acid 5-amide-1,2-13C2) | 56-85-9 | 98.0% | 148.13 | C5H10N2O3 (1,2-13C2) | 10 MG
SDP
Catalog No. 5000295835
Encompass_Preferred
L-Glutamine-1,2-13C2 is an isotopically labeled form of the amino acid L-glutamine supplied as a solid for research use in tracer studies, metabolic flux analysis, and mass spectrometry or NMR experiments.
  • Isotopically labeled at positions 1 and 2 with 13C for tracer experiments.
  • High purity suitable for analytical applications.
  • Supplied in a small, accurately measured quantity for precise dosing.
  • Suitable for mass spectrometry and NMR-based metabolic studies.
  • Solid form with defined molecular weight for stoichiometric calculations.
Catalog No. 50-002-95835 Supplier Medchemexpress LLC Supplier No. HYN0390S1010MG
L-Glutamine-1,2-13C2 is an isotopically labeled form of the amino acid L-glutamine supplied as a solid for research use in tracer studies, metabolic flux analysis, and mass spectrometry or NMR experiments.
  • Isotopically labeled at positions 1 and 2 with 13C for tracer experiments.
  • High purity suitable for analytical applications.
  • Supplied in a small, accurately measured quantity for precise dosing.
  • Suitable for mass spectrometry and NMR-based metabolic studies.
  • Solid form with defined molecular weight for stoichiometric calculations.