Learn More
STEMCELL Technologies L-Glutamine, Size: 100 mL
SDP
Catalog No. NC9535443
Encompass
L-Glutamine (200 mM) is recommended for use as a cell culture supplement and is required for the preparation of MesenCult™-ACF Plus Medium (Catalog #05445).
Catalog No. NC9535443 Supplier STEMCELL Technologies Supplier No. 07100
L-Glutamine (200 mM) is recommended for use as a cell culture supplement and is required for the preparation of MesenCult™-ACF Plus Medium (Catalog #05445).