Learn More
Research Products International Corp L-Glutamine, 100 Grams
SDP
Supplier:  Research Products International Corp G36040100.0
Encompass_Preferred
L-Glutamine is an amino acid essential for various cellular functions in both normal and pathological conditions.
In life science research, it holds significant importance due to its role in cell culture as a key component of cell culture media. L-Glutamine serves as a major energy source for cells, supports protein synthesis, regulates cell proliferation, and plays a critical role in maintaining cellular homeostasis.
Its versatility and indispensability make it a cornerstone in numerous biological studies, ranging from basic cell biology to advanced biomedical research, including cancer biology, immunology, and neuroscience.
Catalog No. 50-213-338