Learn More
Cambridge Isotope Laboratories L-Glutamine 13C5, 184161-19-1, MFCD00144603, 0.1 G
Catalog No. NC0716477
Encompass
Linear Formula: H2N13CO(13CH2)213CH(NH2)13CO2H, Molecular Weight: 151.11, Synonym: L-Glutamic acid 5-amide-13C5, (S)-2,5-Diamino-5-oxopentanoic acid-13C5, L-Glutamine-ul-13C, 13C Labeled glutamine, 99%, MP: 185 °C (dec.) (lit.).
Catalog No. NC0716477 Supplier Cambridge Isotope Laboratories Supplier No. CLM1822H0.1
Linear Formula: H2N13CO(13CH2)213CH(NH2)13CO2H, Molecular Weight: 151.11, Synonym: L-Glutamic acid 5-amide-13C5, (S)-2,5-Diamino-5-oxopentanoic acid-13C5, L-Glutamine-ul-13C, 13C Labeled glutamine, 99%, MP: 185 °C (dec.) (lit.).