Learn More
Medchemexpress LLC L-Glutamine-13C5,15N2 | 285978-14-5 | 153.09 | 1 MG
SDP
Catalog No. 5000388825
Encompass_Preferred
L-Glutamine-13C5,15N2 is the 13C- and 15N-labeled L-Glutamine. It is a non-essential amino acid abundantly present throughout the body and involved in many metabolic processes, providing a source of carbons for oxidation in some cells. This product is for research use only.
  • 13C- and 15N-labeled L-Glutamine
  • Non-essential amino acid
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-88825 Supplier Medchemexpress LLC Supplier No. HYN0390S61MG
L-Glutamine-13C5,15N2 is the 13C- and 15N-labeled L-Glutamine. It is a non-essential amino acid abundantly present throughout the body and involved in many metabolic processes, providing a source of carbons for oxidation in some cells. This product is for research use only.
  • 13C- and 15N-labeled L-Glutamine
  • Non-essential amino acid
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS