Learn More
Medchemexpress LLC L-glutamine-13C5,15N2 | 285978-14-5 | 98.0% | 153.09 g/mol | 13C5H10 15N2 O3 | 10 MG
SDP
Catalog No. 5000296254
Encompass_Preferred
L-Glutamine-13C5,15N2 is an isotopically labeled form of L-glutamine in which five carbon atoms are 13C and two nitrogen atoms are 15N. Supplied as a 10 mg solid with 98.0% purity, it is intended for use as an internal standard, tracer, or NMR reference in metabolic and mass spectrometry studies.
  • Isotopically labeled (13C5, 15N2).
  • High purity: 98.0%.
  • Supplied as a 10 mg solid.
  • Suitable for metabolic tracing and flux analysis.
  • Appropriate as an internal standard for mass spectrometry.
  • Useful for NMR and structural studies.
Catalog No. 50-002-96254 Supplier Medchemexpress LLC Supplier No. HYN0390S610MG
L-Glutamine-13C5,15N2 is an isotopically labeled form of L-glutamine in which five carbon atoms are 13C and two nitrogen atoms are 15N. Supplied as a 10 mg solid with 98.0% purity, it is intended for use as an internal standard, tracer, or NMR reference in metabolic and mass spectrometry studies.
  • Isotopically labeled (13C5, 15N2).
  • High purity: 98.0%.
  • Supplied as a 10 mg solid.
  • Suitable for metabolic tracing and flux analysis.
  • Appropriate as an internal standard for mass spectrometry.
  • Useful for NMR and structural studies.