Learn More
Medchemexpress LLC L-Glutamine-13C5 | 184161-19-1 | 151.11 | 1 ML
SDP
Catalog No. 5000389987
Encompass_Preferred
L-Glutamine-13C5 is the 13C-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid present abundantly throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Involved in many metabolic processes
Catalog No. 50-003-89987 Supplier Medchemexpress LLC Supplier No. HYN0390S110MM/1ML
L-Glutamine-13C5 is the 13C-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid present abundantly throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Involved in many metabolic processes