Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Glutamine-13C5 | 184161-19-1 | MFCD00144603 | 99.9% | 151.11 g/mol | 13C5H10N2O3 | 10 MG
SDP

Catalog No. 5000295504
Encompass_Preferred

L-Glutamine-13C5 is the 13C-labeled form of the amino acid L-glutamine, supplied as a high-purity solid stable isotope for use as a tracer and quantitative standard in metabolic and mass-spectrometry studies.

  • Uniformly labeled with five 13C atoms.
  • Purity 99.9% (typical, rounded).
  • Molecular weight 151.11 g/mol.
  • CAS number 184161-19-1.
  • Physical form 10 mg solid powder.
  • Storage: powder at -20°C for long-term stability.

Catalog No. 50-002-95504 Supplier Medchemexpress LLC Supplier No. HYN0390S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamine-13C5 is the 13C-labeled form of the amino acid L-glutamine, supplied as a high-purity solid stable isotope for use as a tracer and quantitative standard in metabolic and mass-spectrometry studies.

  • Uniformly labeled with five 13C atoms.
  • Purity 99.9% (typical, rounded).
  • Molecular weight 151.11 g/mol.
  • CAS number 184161-19-1.
  • Physical form 10 mg solid powder.
  • Storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.