Learn More
AdipoGen L-Glutamine
SDP
Catalog No. 502560833
Encompass_Preferred
Chemical. CAS 56-85-9. Formula C5H10N2O3. MW 146.14. L-Glutamine is an amino acid and is essential in the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate and other amino acids as well as in protein synthesis and glucose production. It plays also a role in lipid synthesis e.g. in cancer cells, cellular energy as a source next to glucose, nitrogen donation for many anabolic processes, including the synthesis of purines and carbon donation, as a source refilling the citric acid cycle. It is a nontoxic transporter of ammonia in the blood circulation and a precursor to the neurotransmitter glutamate and GABA. L-Glutamine is an essential amino acid which is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine exerts potent antioxidant and anti-inflammatory effects, next to its roles in cell proliferation and cancer.
Catalog No. 50-256-0833 Supplier AdipoGen Supplier No. CDXG0041G025
Chemical. CAS 56-85-9. Formula C5H10N2O3. MW 146.14. L-Glutamine is an amino acid and is essential in the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate and other amino acids as well as in protein synthesis and glucose production. It plays also a role in lipid synthesis e.g. in cancer cells, cellular energy as a source next to glucose, nitrogen donation for many anabolic processes, including the synthesis of purines and carbon donation, as a source refilling the citric acid cycle. It is a nontoxic transporter of ammonia in the blood circulation and a precursor to the neurotransmitter glutamate and GABA. L-Glutamine is an essential amino acid which is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine exerts potent antioxidant and anti-inflammatory effects, next to its roles in cell proliferation and cancer.