Learn More
Medchemexpress LLC L-Glutamine-15N-1 | 59681-32-2 | 147.14 | 25 MG
SDP
Catalog No. 5000389497
Encompass_Preferred
L-Glutamine-15N-1 is the 15N-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid abundantly present throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
Catalog No. 50-003-89497 Supplier Medchemexpress LLC Supplier No. 25MGHYN0390S9
Add to Cart
Add to Cart
L-Glutamine-15N-1 is the 15N-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid abundantly present throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development.