Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamine-15N-1 (L-glutamic acid 5-amide-15N-1) | 59681-32-2 | MFCD00084214 | 98.0% | 147.14 g/mol | C5H10N15NO3 | 10 MG
SDP

Catalog No. 5000295739
Encompass_Preferred

L-Glutamine-15N-1 is the 15N-labeled form of the amino acid L-glutamine used as a stable isotope tracer and analytical standard for metabolic labeling and mass spectrometry applications.

  • Isotope-labeled L-glutamine for tracer and analytical studies.
  • Purity ≥98.0% as stated by the manufacturer.
  • Available as small solid packages (for example 10 mg) and as a 10 mM aqueous solution.
  • Soluble in water (25 mg/mL; ultrasonic may be needed) and in PBS for working solutions.
  • White to off-white solid; powder storage recommended at -20°C for long-term stability.

Catalog No. 50-002-95739 Supplier Medchemexpress LLC Supplier No. HYN0390S910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamine-15N-1 is the 15N-labeled form of the amino acid L-glutamine used as a stable isotope tracer and analytical standard for metabolic labeling and mass spectrometry applications.

  • Isotope-labeled L-glutamine for tracer and analytical studies.
  • Purity ≥98.0% as stated by the manufacturer.
  • Available as small solid packages (for example 10 mg) and as a 10 mM aqueous solution.
  • Soluble in water (25 mg/mL; ultrasonic may be needed) and in PBS for working solutions.
  • White to off-white solid; powder storage recommended at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.